siRNA Details
| Gene Symbol: MROH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207414.2 | NP_997297.2 | 15 | 1915 | ||
| NR_102363.2 | 13 | 1578 | ||||
| NR_102364.2 | 14 | 1849 | ||||
| GenBank mRNA | AK125848.1 | - | 15 | 1915 | ||
| AK131461.1 | - | 14 | 1849 | |||
| BC144560.1 | - | 13 | 1554 | |||

MROH5
Genomic Map