siRNA Details
| Gene Symbol: C9orf172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080482.2 | NP_001073951.2 | 1 | 1135 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9orf172| Gene Symbol: C9orf172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080482.2 | NP_001073951.2 | 1 | 1135 | ||