siRNA Details
Gene Symbol: LOC389834 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027420.1 | 1 | 1113 | |||
GenBank mRNA | AK123403.1 | - | 1 | 1113 | ||
BC137367.1 | - | 1 | 957 | 9021365 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC389834 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027420.1 | 1 | 1113 | |||
GenBank mRNA | AK123403.1 | - | 1 | 1113 | ||
BC137367.1 | - | 1 | 957 | 9021365 |