siRNA Details
Gene Symbol: LOC642249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724939.3 | XP_006725002.1 | 4 | 1735 |
Gene Symbol: LOC389834 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027420.1 | 1 | 1730 | |||
GenBank mRNA | AK123403.1 | - | 1 | 1730 | ||
BC137367.1 | - | 1 | 1574 | 9021365 |
