siRNA Details
| Gene Symbol: PAGE2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015038.2 | NP_001015038.1 | 4 | 275 | ||
| XM_011530785.2 | XP_011529087.1 | 5 | 691 | |||
| XM_011530786.2 | XP_011529088.1 | 4 | 358 | |||
| XM_011530787.2 | XP_011529089.1 | 4 | 338 | |||
| XM_017029513.1 | XP_016885002.1 | 4 | 295 | |||
| GenBank mRNA | BC130359.1 | - | 4 | 234 | 40146390 | |
| BM782405.1 | - | 4 | 260 | |||
| Gene Symbol: PAGE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207339.3 | NP_997222.1 | 4 | 268 | ||
| XM_017029353.1 | XP_016884842.1 | 4 | 297 | |||
| GenBank mRNA | BC054022.1 | - | 4 | 262 | 6603202 | |
| BX113994.1 | - | 4 | 198 | |||

PAGE2B
Genomic Map