siRNA Details
| Gene Symbol: LOC389895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271560.1 | NP_001258489.1 | 1 | 454 | ||
| GenBank mRNA | BM701778.1 | - | 1 | 454 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC389895| Gene Symbol: LOC389895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271560.1 | NP_001258489.1 | 1 | 454 | ||
| GenBank mRNA | BM701778.1 | - | 1 | 454 | ||