siRNA Details
Gene Symbol: LOC389895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271560.1 | NP_001258489.1 | 1 | 480 | ||
GenBank mRNA | BM701778.1 | - | 1 | 480 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC389895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271560.1 | NP_001258489.1 | 1 | 480 | ||
GenBank mRNA | BM701778.1 | - | 1 | 480 |