siRNA Details
| Gene Symbol: OR52K1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005171.2 | NP_001005171.2 | 1 | 607 | ||
| GenBank mRNA | BC137056.1 | - | 1 | 607 | 9021053 | |
| BC137059.1 | - | 1 | 607 | 9021056 | ||
| Gene Symbol: OR52K2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005172.2 | NP_001005172.2 | 1 | 630 | ||
| GenBank mRNA | BC137247.1 | - | 1 | 630 | 9021244 | |
| BC137248.1 | - | 1 | 630 | 9021245 | ||

OR52K1
Genomic Map