siRNA Details
| Gene Symbol: COL6A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004369.3 | NP_004360.2 | 6 | 2654 | ||
| NM_057164.4 | NP_476505.3 | 4 | 1433 | |||
| NM_057165.4 | NP_476506.3 | 5 | 2036 | |||
| NM_057167.3 | NP_476508.2 | 5 | 2036 | |||
| XM_005246066.1 | XP_005246123.1 | 3 | 1157 | |||
| XM_006712253.1 | XP_006712316.1 | 6 | 2611 | |||
| XM_011510574.1 | XP_011508876.1 | 6 | 2611 | |||
| XM_017003303.1 | XP_016858792.1 | 11 | 2661 | |||
| GenBank mRNA | AK092021.1 | - | 4 | 1359 | ||
| AK304870.1 | - | 5 | 1971 | |||
| BX647500.1 | - | 5 | 1998 | |||
| X52022.1 | - | 6 | 2624 | |||

COL6A3
Genomic Map