siRNA Details
| Gene Symbol: OR52A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005160.2 | NP_001005160.1 | 1 | 714 | ||
| GenBank mRNA | AY726590.1 | - | 1 | 650 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR52A5| Gene Symbol: OR52A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005160.2 | NP_001005160.1 | 1 | 714 | ||
| GenBank mRNA | AY726590.1 | - | 1 | 650 | ||