siRNA Details
Gene Symbol: FLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002016.1 | NP_002007.1 | 2 | 11977 | ||
GenBank mRNA | M60502.1 | - | 3 | 2149 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002016.1 | NP_002007.1 | 2 | 11977 | ||
GenBank mRNA | M60502.1 | - | 3 | 2149 |