siRNA Details
| Gene Symbol: OR51I2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004754.2 | NP_001004754.1 | 1 | 665 | ||
| GenBank mRNA | BC140730.1 | - | 1 | 665 | 9021598 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR51I2| Gene Symbol: OR51I2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004754.2 | NP_001004754.1 | 1 | 665 | ||
| GenBank mRNA | BC140730.1 | - | 1 | 665 | 9021598 | |