siRNA Details
Gene Symbol: OR52D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC136928.1 | - | 1 | 424 | 9020920 | |
BC136930.1 | - | 1 | 424 | 9020922 |

More Information
Other Species Targeted


Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR52D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC136928.1 | - | 1 | 424 | 9020920 | |
BC136930.1 | - | 1 | 424 | 9020922 |