siRNA Details
| Gene Symbol: OR52E8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005168.1 | NP_001005168.1 | 1 | 880 | ||
| GenBank mRNA | BC137030.1 | - | 1 | 912 | 9021026 | |
| BC137041.1 | - | 1 | 912 | 9021037 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR52E8| Gene Symbol: OR52E8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005168.1 | NP_001005168.1 | 1 | 880 | ||
| GenBank mRNA | BC137030.1 | - | 1 | 912 | 9021026 | |
| BC137041.1 | - | 1 | 912 | 9021037 | ||