siRNA Details
| Gene Symbol: OR52E4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005165.1 | NP_001005165.1 | 1 | 424 | ||
| GenBank mRNA | BC136972.1 | - | 1 | 446 | 9020964 | |
| BC136974.1 | - | 1 | 446 | 9020966 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR52E4| Gene Symbol: OR52E4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005165.1 | NP_001005165.1 | 1 | 424 | ||
| GenBank mRNA | BC136972.1 | - | 1 | 446 | 9020964 | |
| BC136974.1 | - | 1 | 446 | 9020966 | ||