siRNA Details
| Gene Symbol: PRKCSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001329.2 | NP_001001329.1 | 14 | 1714 | ||
| NM_001289102.1 | NP_001276031.1 | 13 | 1565 | |||
| NM_001289103.1 | NP_001276032.1 | 14 | 1748 | |||
| NM_001289104.1 | NP_001276033.1 | 14 | 1744 | |||
| NM_002743.3 | NP_002734.2 | 14 | 1335 | |||
| XM_011528130.1 | XP_011526432.1 | 15 | 1510 | |||
| XM_011528131.1 | XP_011526433.1 | 15 | 1489 | |||
| XM_011528132.1 | XP_011526434.1 | 15 | 1480 | |||
| XM_017026977.1 | XP_016882466.1 | 14 | 1339 | |||
| GenBank mRNA | AF144075.1 | - | 13 | 1196 | ||
| AK130663.1 | - | 14 | 1342 | |||
| AK225279.1 | - | 14 | 1364 | |||
| AK225390.1 | - | 14 | 1325 | |||
| AK290433.1 | - | 13 | 1560 | |||
| AK296187.1 | - | 14 | 1371 | |||
| BC013586.1 | - | 13 | 1130 | |||
| BC015154.1 | - | 4 | 212 | |||
| BT009858.1 | - | 9 | 806 | |||
| CF618983.1 | - | 4 | 239 | |||
| J03075.1 | - | 14 | 1329 | |||
| KU178278.1 | - | 9 | 809 | |||
| KU178279.1 | - | 9 | 830 | |||

PRKCSH
Genomic Map