siRNA Details
| Gene Symbol: OR8H2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005200.1 | NP_001005200.1 | 1 | 239 | ||
| Gene Symbol: OR8H3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005201.1 | NP_001005201.1 | 1 | 239 | ||

OR8H2
Genomic Map