siRNA Details
| Gene Symbol: OR8K1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002907.1 | NP_001002907.1 | 1 | 444 | ||
| GenBank mRNA | BC146844.1 | - | 1 | 538 | 9056772 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR8K1| Gene Symbol: OR8K1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002907.1 | NP_001002907.1 | 1 | 444 | ||
| GenBank mRNA | BC146844.1 | - | 1 | 538 | 9056772 | |