siRNA Details
| Gene Symbol: OR5M10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004741.1 | NP_001004741.1 | 1 | 866 | ||
| GenBank mRNA | BC140733.1 | - | 1 | 932 | 9021601 | |
| Gene Symbol: OR5M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004740.1 | NP_001004740.1 | 1 | 866 | ||

OR5M10
Genomic Map