siRNA Details
| Gene Symbol: OR9G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005213.1 | NP_001005213.1 | 1 | 739 | ||
| Gene Symbol: OR9G9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013358.2 | NP_001013376.2 | 1 | 739 | ||
More Information
Set Membership: |
|

OR9G1
Genomic Map