siRNA Details
| Gene Symbol: OR5B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005566.2 | NP_001005566.1 | 1 | 107 | ||
| GenBank mRNA | BC136922.1 | - | 1 | 110 | 9020914 | |
| BC136923.1 | - | 1 | 104 | 9020915 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR5B2| Gene Symbol: OR5B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005566.2 | NP_001005566.1 | 1 | 107 | ||
| GenBank mRNA | BC136922.1 | - | 1 | 110 | 9020914 | |
| BC136923.1 | - | 1 | 104 | 9020915 | ||