siRNA Details
| Gene Symbol: OR5B12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004733.2 | NP_001004733.1 | 1 | 782 | ||
| GenBank mRNA | BC136947.1 | - | 1 | 782 | 9020939 | |
| BC136948.1 | - | 1 | 782 | 9020940 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR5B12| Gene Symbol: OR5B12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004733.2 | NP_001004733.1 | 1 | 782 | ||
| GenBank mRNA | BC136947.1 | - | 1 | 782 | 9020939 | |
| BC136948.1 | - | 1 | 782 | 9020940 | ||