siRNA Details
| Gene Symbol: MLANA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005511.1 | NP_005502.1 | 3, 4 | 235 | ||
| GenBank mRNA | AK312149.1 | - | 3, 4 | 251 | ||
| BC014423.1 | - | 3, 4 | 260 | 4639927 | ||
| CR450299.1 | - | 2, 3 | 182 | |||
| U06452.1 | - | 3, 4 | 235 | |||
| U06654.1 | - | 3, 4 | 342 | |||

MLANA
Genomic Map