siRNA Details
| Gene Symbol: OR4D10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004705.1 | NP_001004705.1 | 1 | 194 | ||
| GenBank mRNA | BC136893.1 | - | 1 | 251 | 9020884 | |
| BC136894.1 | - | 1 | 251 | 9020885 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR4D10| Gene Symbol: OR4D10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004705.1 | NP_001004705.1 | 1 | 194 | ||
| GenBank mRNA | BC136893.1 | - | 1 | 251 | 9020884 | |
| BC136894.1 | - | 1 | 251 | 9020885 | ||