siRNA Details
| Gene Symbol: LEMD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167614.1 | NP_001161086.1 | 9, 10 | 2336 | ||
| NM_014319.4 | NP_055134.2 | 9, 10 | 2339 | |||
| GenBank mRNA | AF112299.2 | - | 9, 10 | 2319 | ||
| AK024954.1 | - | 9, 10 | 1014 | |||
| AK295406.1 | - | 3, 4 | 387 | |||
| AK312473.1 | - | 9, 10 | 2336 | |||
| AL137533.1 | - | 1, 2 | 175 | |||

LEMD3
Genomic Map