siRNA Details
| Gene Symbol: LEMD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167614.1 | NP_001161086.1 | 12, 13 | 2604 | ||
| NM_014319.4 | NP_055134.2 | 12, 13 | 2607 | |||
| GenBank mRNA | AF112299.2 | - | 12, 13 | 2587 | ||
| AK024954.1 | - | 12, 13 | 1282 | |||
| AK295406.1 | - | 6, 7 | 655 | |||
| AK312473.1 | - | 12, 13 | 2604 | |||
| AL137533.1 | - | 4, 5 | 443 | |||

LEMD3
Genomic Map