siRNA Details
| Gene Symbol: LEMD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167614.1 | NP_001161086.1 | 1 | 1330 | ||
| NM_014319.4 | NP_055134.2 | 1 | 1330 | |||
| GenBank mRNA | AF112299.2 | - | 1 | 1310 | ||
| AK312473.1 | - | 1 | 1330 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LEMD3| Gene Symbol: LEMD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167614.1 | NP_001161086.1 | 1 | 1330 | ||
| NM_014319.4 | NP_055134.2 | 1 | 1330 | |||
| GenBank mRNA | AF112299.2 | - | 1 | 1310 | ||
| AK312473.1 | - | 1 | 1330 | |||