siRNA Details
| Gene Symbol: OR8B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005467.1 | NP_001005467.1 | 1 | 275 | ||
| XM_017017716.1 | XP_016873205.1 | 6 | 1641 | |||
| XM_017017717.1 | XP_016873206.1 | 6 | 944 | |||
| XM_017017718.1 | XP_016873207.1 | 4 | 1372 | |||

OR8B3
Genomic Map