siRNA Details
| Gene Symbol: OR8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005194.1 | NP_001005194.1 | 1 | 157 | ||
| GenBank mRNA | AY726592.1 | - | 1 | 33 | ||
| BC137194.1 | - | 1 | 229 | 9021191 | ||
| BC137197.1 | - | 1 | 229 | 9021194 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR8A1| Gene Symbol: OR8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005194.1 | NP_001005194.1 | 1 | 157 | ||
| GenBank mRNA | AY726592.1 | - | 1 | 33 | ||
| BC137194.1 | - | 1 | 229 | 9021191 | ||
| BC137197.1 | - | 1 | 229 | 9021194 | ||