siRNA Details
| Gene Symbol: NDUFA12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258338.1 | NP_001245267.1 | 3 | 388 | ||
| NM_018838.4 | NP_061326.1 | 4 | 476 | |||
| GenBank mRNA | AF112208.1 | - | 4 | 496 | ||
| AF217092.1 | - | 4 | 455 | |||
| AK222619.1 | - | 4 | 463 | |||
| BC005936.1 | - | 4 | 465 | 4069306 | ||
| BG943528.1 | - | 4 | 461 | |||
| BM548072.1 | - | 543 | ||||
| BU532909.1 | - | 3 | 375 | |||
| U34343.1 | - | 582 | ||||

NDUFA12
Genomic Map