siRNA Details
| Gene Symbol: PCDH19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105243.1 | NP_001098713.1 | 4, 5 | 4384 | ||
| NM_001184880.1 | NP_001171809.1 | 5, 6 | 4525 | |||
| NM_020766.2 | NP_065817.2 | 4, 5 | 4381 | |||
| XM_011530997.2 | XP_011529299.1 | 5, 6 | 3491 | |||
| GenBank mRNA | AB037734.1 | - | 4, 5 | 599 | ||
| AK096591.1 | - | 4, 5 | 1337 | |||
| BC136628.1 | - | 4, 5 | 1265 | 9020618 | ||
| CR749278.1 | - | 4, 5 | 1262 | |||
| EF676096.1 | - | 5, 6 | 4525 | |||

PCDH19
Genomic Map