siRNA Details
| Gene Symbol: ROBO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022370.3 | NP_071765.2 | 14 | 2324 | ||
| XM_017018122.1 | XP_016873611.1 | 13 | 2089 | |||
| GenBank mRNA | AK291045.1 | - | 14 | 2258 | ||
| AY509035.1 | - | 14 | 2347 | |||

ROBO3
Genomic Map