siRNA Details
| Gene Symbol: RNASE9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001673.3 | NP_001001673.2 | 2 | 402 | ||
| NM_001110356.1 | NP_001103826.1 | 3 | 454 | |||
| NM_001110357.1 | NP_001103827.1 | 3 | 437 | |||
| NM_001110358.1 | NP_001103828.1 | 4 | 490 | |||
| NM_001110359.1 | NP_001103829.1 | 5 | 600 | |||
| NM_001110360.1 | NP_001103830.1 | 4 | 565 | |||
| NM_001110361.1 | NP_001103831.1 | 5 | 525 | |||
| NM_001289110.1 | NP_001276039.1 | 4 | 489 | |||
| GenBank mRNA | AF382949.1 | - | 1 | 330 | ||
| AY665804.1 | - | 1 | 330 | |||
| AY907670.1 | - | 2 | 402 | |||
| BC130311.1 | - | 1 | 415 | 40146342 | ||
| BC130313.1 | - | 1 | 415 | 40146344 | ||
| EF061296.1 | - | 3 | 437 | |||
| EF061297.1 | - | 3 | 454 | |||
| EF061298.1 | - | 4 | 489 | |||
| EF061299.1 | - | 4 | 490 | |||
| EF061300.1 | - | 4 | 565 | |||
| EF061301.1 | - | 5 | 525 | |||
| EF061302.1 | - | 5 | 600 | |||
| EF061303.1 | - | 3 | 2021 | |||
| EF061304.1 | - | 4 | 2056 | |||
| EU414264.1 | - | 1 | 330 | |||

RNASE9
Genomic Map