siRNA Details
| Gene Symbol: UBAP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163692.1 | NP_001157164.1 | 2 | 427 | ||
| XM_011521547.2 | XP_011519849.1 | 2 | 724 | |||
| XM_017022172.1 | XP_016877661.1 | 2 | 724 | |||
| GenBank mRNA | BQ677048.1 | - | 3 | 525 | ||

UBAP1L
Genomic Map