siRNA Details
| Gene Symbol: UBAP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163692.1 | NP_001157164.1 | 3 | 962 | ||
| XM_011521547.2 | XP_011519849.1 | 3 | 1259 | |||
| XM_017022172.1 | XP_016877661.1 | 3 | 1259 | |||
| GenBank mRNA | CR749500.1 | - | 1 | 2034 | ||

UBAP1L
Genomic Map