siRNA Details
| Gene Symbol: KRT39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213656.3 | NP_998821.3 | 1 | 316 | ||
| GenBank mRNA | AJ786657.1 | - | 1 | 316 | ||
| BC157887.1 | - | 1 | 310 | 8862708 | ||
| BC171907.1 | - | 1 | 310 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT39| Gene Symbol: KRT39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213656.3 | NP_998821.3 | 1 | 316 | ||
| GenBank mRNA | AJ786657.1 | - | 1 | 316 | ||
| BC157887.1 | - | 1 | 310 | 8862708 | ||
| BC171907.1 | - | 1 | 310 | |||