siRNA Details
| Gene Symbol: FAH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000137.2 | NP_000128.1 | 11, 12 | 1116 | ||
| GenBank mRNA | AK313951.1 | - | 11, 12 | 1111 | ||
| BC002527.1 | - | 11, 12 | 954 | 3139988 | ||
| BT007160.1 | - | 11, 12 | 952 | |||
| BX537608.1 | - | 10, 11 | 2981 | |||
| M55150.1 | - | 11, 12 | 1008 | |||
| S63548.1 | - | 1, 2 | 50 | |||
| X51728.1 | - | 10, 11 | 879 | |||

FAH
Genomic Map