siRNA Details
| Gene Symbol: HGD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000187.3 | NP_000178.2 | 13 | 1507 | ||
| XM_005247412.1 | XP_005247469.1 | 11 | 1282 | |||
| XM_017006277.1 | XP_016861766.1 | 13 | 1120 | |||
| GenBank mRNA | AF045167.1 | - | 13 | 1214 | ||
| AK124291.1 | - | 8 | 1891 | |||
| AK290782.1 | - | 13 | 1213 | |||
| AK313563.1 | - | 13 | 1491 | |||
| BC071757.1 | - | 13 | 1145 | 6714217 | ||
| U63008.1 | - | 13 | 1176 | |||
| Z75048.1 | - | 13 | 1176 | |||

HGD
Genomic Map