siRNA Details
| Gene Symbol: NPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005258277.1 | XP_005258334.1 | 25 | 4045 | ||
| XM_005258279.2 | XP_005258336.1 | 25 | 3994 | |||
| XM_011526015.1 | XP_011524317.1 | 25 | 4293 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NPC1| Gene Symbol: NPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005258277.1 | XP_005258334.1 | 25 | 4045 | ||
| XM_005258279.2 | XP_005258336.1 | 25 | 3994 | |||
| XM_011526015.1 | XP_011524317.1 | 25 | 4293 | |||