siRNA Details
| Gene Symbol: ADAMTS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014244.4 | NP_055059.2 | 7 | 1282 | ||
| NM_021599.3 | NP_067610.1 | 7 | 1282 | |||
| XM_017010063.1 | XP_016865552.1 | 6 | 718 | |||
| GenBank mRNA | AJ003125.1 | - | 7 | 1180 | ||
| AK308213.1 | - | 7 | 1285 | |||

ADAMTS2
Genomic Map