siRNA Details
| Gene Symbol: CLN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017882.2 | NP_060352.1 | 7 | 1615 | ||
| GenBank mRNA | AK000568.1 | - | 7 | 1556 | ||
| AK027604.1 | - | 7 | 1579 | |||
| AK074753.1 | - | 4 | 1105 | |||
| AK291175.1 | - | 7 | 1538 | |||
| BC010849.1 | - | 7 | 1525 | 3878776 | ||
| BC013130.1 | - | 7 | 1462 | 4517748 | ||
| CN427564.1 | - | 1 | 381 | |||

CLN6
Genomic Map