siRNA Details
| Gene Symbol: NAGS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153006.2 | NP_694551.1 | 4 | 1076 | ||
| XM_011524438.1 | XP_011522740.1 | 4 | 1194 | |||
| XM_011524439.2 | XP_011522741.1 | 4 | 966 | |||
| GenBank mRNA | AK074407.1 | - | 4 | 982 | ||
| AK091938.1 | - | 2 | 351 | |||
| AK300923.1 | - | 5 | 982 | |||
| AK314432.1 | - | 4 | 1194 | |||
| AY116538.1 | - | 4 | 1076 | |||
| AY158070.1 | - | 4 | 797 | |||
| BC111713.1 | - | 5 | 862 | 40005903 | ||

NAGS
Genomic Map