siRNA Details
| Gene Symbol: ACP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004318.2 | NP_001004318.2 | 3 | 545 | ||
| XM_011526966.2 | XP_011525268.1 | 3 | 597 | |||
| XM_011526967.2 | XP_011525269.1 | 3 | 545 | |||
| XM_011526969.1 | XP_011525271.1 | 3 | 545 | |||
| XM_017026807.1 | XP_016882296.1 | 3 | 1396 | |||
| XM_017026808.1 | XP_016882297.1 | 3 | 582 | |||
| GenBank mRNA | AK131245.1 | - | 3 | 545 | ||
| BC136721.1 | - | 2 | 347 | 9020711 | ||
| BC136722.1 | - | 2 | 347 | 9020712 | ||
| BC142613.1 | - | 3 | 494 | |||

ACP7
Genomic Map