siRNA Details
| Gene Symbol: PINLYP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193621.1 | NP_001180550.1 | 5 | 549 | ||
| NM_001193622.1 | NP_001180551.1 | 3 | 209 | |||
| NM_001321124.1 | NP_001308053.1 | 3 | 229 | |||
| XM_011526970.2 | XP_011525272.1 | 4 | 2145 | |||
| XM_011526971.2 | XP_011525273.1 | 3 | 571 | |||
| GenBank mRNA | AK129550.1 | - | 3 | 197 | ||
| BC071811.1 | - | 3 | 209 | |||
| CN355575.1 | - | 3 | 188 | |||
| HY005068.1 | - | 3 | 229 | |||

PINLYP
Genomic Map