siRNA Details
| Gene Symbol: PEX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002617.3 | NP_002608.1 | 3 | 315 | ||
| NM_153818.1 | NP_722540.1 | 3 | 315 | |||
| XM_011541573.1 | XP_011539875.1 | 3 | 366 | |||
| XM_011541576.1 | XP_011539878.1 | 3 | 366 | |||
| GenBank mRNA | AB013818.1 | - | 3 | 315 | ||
| AF060502.1 | - | 3 | 278 | |||
| AK124816.1 | - | 3 | 293 | |||
| BC000543.2 | - | 3 | 274 | 3163782 | ||
| BC018198.1 | - | 3 | 297 | 3882695 | ||
| CR542077.1 | - | 3 | 246 | |||
| DA439481.1 | - | 3 | 316 | |||

PEX10
Genomic Map