siRNA Details
| Gene Symbol: PEX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002617.3 | NP_002608.1 | 3 | 632 | ||
| NM_153818.1 | NP_722540.1 | 3 | 632 | |||
| XM_011541573.1 | XP_011539875.1 | 3 | 683 | |||
| XM_011541576.1 | XP_011539878.1 | 3 | 683 | |||
| GenBank mRNA | AB013818.1 | - | 3 | 632 | ||
| AF060502.1 | - | 3 | 595 | |||
| AK124816.1 | - | 3 | 610 | |||
| BC000543.2 | - | 3 | 591 | 3163782 | ||
| BC018198.1 | - | 3 | 614 | 3882695 | ||
| CR542077.1 | - | 3 | 563 | |||

PEX10
Genomic Map