siRNA Details
| Gene Symbol: NDUFS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004553.4 | NP_004544.1 | 4 | 508 | ||
| GenBank mRNA | AF044959.1 | - | 4 | 482 | ||
| AK022503.1 | - | 4 | 491 | |||
| BC038664.1 | - | 4 | 487 | 5204222 | ||
| BC046155.2 | - | 4 | 477 | 6047460 | ||
| BG706812.1 | - | 4 | 497 | |||
| BM172493.1 | - | 4 | 363 | |||
| BM759028.1 | - | 4 | 495 | |||
| BM764340.1 | - | 4 | 495 | |||
| CN285188.1 | - | 4 | 462 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ674245.1 | - | 460 | Chr. - - [-] on Build | ||

NDUFS6
Genomic Map