siRNA Details
| Gene Symbol: SPATA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040428.3 | NP_001035518.1 | 7 | 1063 | ||
| NM_018418.4 | NP_060888.2 | 8 | 1159 | |||
| XM_005267851.1 | XP_005267908.1 | 8 | 1133 | |||
| XM_005267852.1 | XP_005267909.1 | 7 | 1062 | |||
| XM_005267854.1 | XP_005267911.1 | 9 | 1195 | |||
| XM_005267855.1 | XP_005267912.1 | 8 | 1128 | |||
| XM_006720204.1 | XP_006720267.1 | 8 | 1133 | |||
| XM_006720205.1 | XP_006720268.1 | 8 | 1133 | |||
| XM_011536951.1 | XP_011535253.1 | 8 | 1327 | |||
| XM_011536952.1 | XP_011535254.1 | 6 | 945 | |||
| XM_011536953.1 | XP_011535255.1 | 7 | 1266 | |||
| XM_017021452.1 | XP_016876941.1 | 8 | 1324 | |||
| XM_017021453.1 | XP_016876942.1 | 8 | 1293 | |||
| XM_017021454.1 | XP_016876943.1 | 9 | 1192 | |||
| XM_017021455.1 | XP_016876944.1 | 8 | 1125 | |||
| XM_017021456.1 | XP_016876945.1 | 9 | 1138 | |||
| XM_017021457.1 | XP_016876946.1 | 7 | 1263 | |||
| GenBank mRNA | AF144487.3 | - | 8 | 1139 | ||
| AF144488.2 | - | 6 | 867 | |||
| AK055864.1 | - | 7 | 2731 | |||
| AK225746.1 | - | 9 | 1133 | |||
| AK290631.1 | - | 7 | 1034 | |||
| AL136604.1 | - | 7 | 1043 | |||
| BC008656.1 | - | 3 | 591 | |||
| BC034635.1 | - | 2226 | ||||
| BC090875.1 | - | 8 | 1446 | 30383306 | ||
| CR533563.1 | - | 7 | 888 | |||
| CR936777.1 | - | 8 | 1105 | |||
| EU668353.1 | - | 8 | 1135 | |||

SPATA7
Genomic Map