siRNA Details
| Gene Symbol: CDH23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171930.1 | NP_001165401.1 | 3 | 496 | ||
| NM_001171931.1 | NP_001165402.1 | 3 | 496 | |||
| NM_001171932.1 | NP_001165403.1 | 3 | 496 | |||
| NM_022124.5 | NP_071407.4 | 3 | 496 | |||
| NM_052836.3 | NP_443068.1 | 3 | 496 | |||
| XM_006717940.3 | XP_006718003.1 | 3 | 490 | |||
| XM_006717942.3 | XP_006718005.1 | 3 | 490 | |||
| XM_011540039.2 | XP_011538341.1 | 3 | 490 | |||
| XM_011540042.2 | XP_011538344.1 | 3 | 490 | |||
| XM_011540043.2 | XP_011538345.1 | 3 | 490 | |||
| XM_011540044.2 | XP_011538346.1 | 3 | 131 | |||
| XM_011540045.2 | XP_011538347.1 | 3 | 490 | |||
| XM_011540048.2 | XP_011538350.1 | 3 | 490 | |||
| XM_011540049.2 | XP_011538351.1 | 3 | 490 | |||
| XM_011540051.2 | XP_011538353.1 | 3 | 490 | |||
| XM_017016499.1 | XP_016871988.1 | 3 | 496 | |||
| XM_017016500.1 | XP_016871989.1 | 3 | 490 | |||
| GenBank mRNA | AF312024.1 | - | 2 | 106 | ||
| AY010111.2 | - | 2 | 106 | |||
| AY358617.1 | - | 3 | 146 | |||
| BC011570.1 | - | 3 | 501 | |||
| BC032581.1 | - | 3 | 502 | |||
| BC065284.1 | - | 3 | 473 | |||
| BC136976.1 | - | 3 | 366 | |||
| BC136977.1 | - | 3 | 365 | |||
| BC139903.1 | - | 3 | 488 | |||

CDH23
Genomic Map