siRNA Details
| Gene Symbol: HGSNAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152419.2 | NP_689632.2 | 4 | 526 | ||
| XM_005273409.1 | XP_005273466.1 | 4 | 526 | |||
| XM_005273410.1 | XP_005273467.1 | 4 | 526 | |||
| XM_005273411.1 | XP_005273468.1 | 4 | 526 | |||
| XM_005273412.3 | XP_005273469.1 | 4 | 526 | |||
| XM_017013088.1 | XP_016868577.1 | 4 | 526 | |||
| XM_017013089.1 | XP_016868578.1 | 4 | 430 | |||
| GenBank mRNA | AK304441.1 | - | 4 | 478 | ||
| BC042037.1 | - | 4 | 471 | |||
| BE327420.1 | - | 3 | 374 | |||
| DA395558.1 | - | 4 | 480 | |||

HGSNAT
Genomic Map